4-Methoxybenzylamine ≥97.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

M0870-25ML M0870-250ML
TCIAM0870-25MLEA 38.5 EUR
TCIAM0870-25ML TCIAM0870-250ML
4-Methoxybenzylamine ≥97.0% (by GC, titration analysis)
4-Methoxybenzylamine
Formule: C₈H₁₁NO
Kookpunt: 236°C
Dichtheid: 1,06 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008122
CAS nummer: 2393-23-9
EINECS: 219-247-2
UN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46b5dd34645167b4584890b1468d93d839336962&vid=46b5dd34645167b4584890b1468d93d839336962&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR