2,3,4-Trifluorobenzyl bromide ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 1-(Bromomethyl)-2,3,4-trifluorobenzene, α-Bromo-2,3,4-trifluorotoluene
T2462-1G T2462-5G
TCIAT2462-1GEA 29.7 EUR
TCIAT2462-1G TCIAT2462-5G
2,3,4-Trifluorobenzyl bromide ≥98.0% (by GC)
2,3,4-Trifluorobenzyl bromide
Formule: C₇H₄BrF₃
Kookpunt: 182 °C (1013 hPa)
Smeltpunt: 21 °C
Dichtheid: 1,71 g/cm³ (20 °C)
Vlampunt: 91°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00061233
CAS nummer: 157911-55-2
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=819c7fc837c503aaf253445a72866992522ab0e4&vid=819c7fc837c503aaf253445a72866992522ab0e4&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %