3,4,5-Trifluorobenzyl bromide ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 5-(Bromomethyl)-1,2,3-trifluorobenzene, α-Bromo-3,4,5-trifluorotoluene
T2464-1G T2464-5G
TCIAT2464-1GEA 23.1 EUR
TCIAT2464-1G TCIAT2464-5G
3,4,5-Trifluorobenzyl bromide ≥97.0% (by GC)
3,4,5-Trifluorobenzyl bromide
Formule: C₇H₄BrF₃
Kookpunt: 44 - 46 °C (1 -rr)
Dichtheid: 1,689 g/cm³ (20 °C)
Vlampunt: 65°C
MDL Nummer: MFCD00083528
CAS nummer: 220141-72-0
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=251e45d12dc4d89b0f3b6ca185c6550a578f94a9&vid=251e45d12dc4d89b0f3b6ca185c6550a578f94a9&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light yellow to Brown clear liquid
Purity (GC) min. 97.0 %