α-Bromo-4-iodotoulene ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 4-Iodobenzyl bromide, 1-(Bromomethyl)-4-iodobenzene, α-Bromo-p-iodotoulene

I0571-25G I0571-5G
TCIAI0571-25GEA 215 EUR
TCIAI0571-25G TCIAI0571-5G
α-Bromo-4-iodotoulene ≥97.0% (by GC)
α-Bromo-4-iodotoulene
Formule: C₇H₆BrI
Kookpunt: 115 - 120 °C (0,6 -rr)
Smeltpunt: 80 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00209656
CAS nummer: 16004-15-2
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=acb9d7eedeb79626f61d20b3f1f78f02472d5bee&vid=acb9d7eedeb79626f61d20b3f1f78f02472d5bee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 78.0 to 82.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR