2-Bromo-5-fluoroaniline ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B2163-25G B2163-5G
TCIAB2163-25GEA 73.7 EUR
TCIAB2163-25G TCIAB2163-5G
2-Bromo-5-fluoroaniline ≥98.0% (by GC)
2-Bromo-5-fluoroaniline
Formule: C₆H₅BrFN
Kookpunt: 154°C
MDL Nummer: MFCD00070750
CAS nummer: 1003-99-2
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab1b7a83e56396f6f75da2470c70e10f8901f2ff&vid=ab1b7a83e56396f6f75da2470c70e10f8901f2ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 43.0 to 47.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR