2-Butylaniline ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B1766-25G
B1766-5G
TCIAB1766-25GEA
493
EUR
TCIAB1766-25G
TCIAB1766-5G
2-Butylaniline ≥98.0% (by titrimetric analysis)
2-Butylaniline
Formule:
C₁₀H₁₅N MW: 149.24 g/mol Kookpunt: 126°C |
MDL Nummer:
MFCD00130033 CAS nummer: 2696-85-7 EINECS: 220-272-6 |
|
Specification Test Results
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...