2-Amino-4-chlorophenol ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

A0284-25G A0284-500G
TCIAA0284-25GEA 25.3 EUR
TCIAA0284-25G TCIAA0284-500G
2-Amino-4-chlorophenol ≥98.0% (by GC, titration analysis)
2-Amino-4-chlorophenol
Formule: C₆H₆ClNO
Smeltpunt: 140…143 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007694
CAS nummer: 95-85-2
EINECS: 202-458-9
UN: 2673
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6662ebba3635f1c0238e0fe5a3a9ee853d6e04f3&vid=6662ebba3635f1c0238e0fe5a3a9ee853d6e04f3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 137.0 to 141.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR