3-Fluoro-p-toluidine ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
F0440-25G F0440-5G
TCIAF0440-25GEA 79.2 EUR
TCIAF0440-25G TCIAF0440-5G
3-Fluoro-p-toluidine ≥98.0% (by GC)
3-Fluoro-p-toluidine
Formule: C₇H₈FN
Kookpunt: 97 °C (15 -rr)
Smeltpunt: 37 °C
Dichtheid: 1,09 g/cm³ (20 °C)
MDL Nummer: MFCD00007762
CAS nummer: 452-77-7
EINECS: 207-212-4
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f42df13c9eb29645c8e5f62eec55655d0680dfb7&vid=f42df13c9eb29645c8e5f62eec55655d0680dfb7&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light yellow to Brown powder to lump
Purity (GC) min. 98.0 %
Freezing point 34.0 to 37.0 deg-C
Specificaties
Environmentally Preferable