4-Bromo-3,5-difluoroaniline ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B3416-25G B3416-5G
TCIAB3416-25GEA 192 EUR
TCIAB3416-25G TCIAB3416-5G
4-Bromo-3,5-difluoroaniline ≥98.0% (by GC, titration analysis)
4-Bromo-3,5-difluoroaniline
Formule: C₆H₄BrF₂N
MW: 208.01 g/mol
MDL Nummer: MFCD00236593
CAS nummer: 203302-95-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a3da37a854604934195684970537d15527febdd&vid=5a3da37a854604934195684970537d15527febdd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light gray to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 97.0 to 102.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR