4-Bromo-3-fluoroaniline ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B2102-1G B2102-5G
TCIAB2102-1GEA 17.6 EUR
TCIAB2102-1G TCIAB2102-5G
4-Bromo-3-fluoroaniline ≥98.0% (by GC)
4-Bromo-3-fluoroaniline
Formule: C₆H₅BrFN
Kookpunt: 231 °C (1013 hPa)
Smeltpunt: 72…73 °C
MDL Nummer: MFCD00672933
CAS nummer: 656-65-5
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b54c286598ace309a5af57a425aed166d8d6a0cd&vid=b54c286598ace309a5af57a425aed166d8d6a0cd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 66.0 to 73.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR