2-Amino-5-fluorfenol ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
A2300-1G
A2300-5G
TCIAA2300-1GEA
50.6
EUR
TCIAA2300-1G
TCIAA2300-5G
2-Amino-5-fluorfenol ≥97.0% (by GC)
2-Amino-5-fluorfenol
Formule:
C₆H₆FNO |
MDL Nummer:
MFCD00671759 CAS nummer: 53981-24-1 |
|
Specification Test Results
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...