4-Amino-2-nitrophenol ≥95.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

A0379-25G A0379-5G
TCIAA0379-25GEA 92.4 EUR
TCIAA0379-25G TCIAA0379-5G
4-Amino-2-nitrophenol ≥95.0% (by titrimetric analysis)
4-Amino-2-nitrophenol
Formule: C₆H₆N₂O₃
Smeltpunt: 124…128 °C
MDL Nummer: MFCD00007876
CAS nummer: 119-34-6
EINECS: 204-316-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c65188ca234e70f2d01462ecad47036dc80427a&vid=5c65188ca234e70f2d01462ecad47036dc80427a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Orange to Brown to Dark red powder to crystal
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 127.0 to 130.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR