4-Aminothiophenol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

A0623-25G A0623-5G A0623-100G
TCIAA0623-25GEA 85.8 EUR
TCIAA0623-25G TCIAA0623-5G TCIAA0623-100G
4-Aminothiophenol ≥98.0% (by GC)
4-Aminothiophenol
Formule: C₆H₇NS
Kookpunt: 142°C
Smeltpunt: 40…44 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00007880
CAS nummer: 1193-02-8
EINECS: 214-763-4
UN: 3263
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18a673105207e0bd05f516ff1f37a1f86e191fc6&vid=18a673105207e0bd05f516ff1f37a1f86e191fc6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR