4-Bromo-2,5-dichlorophenol ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
B2822-25G B2822-500G
TCIAB2822-25GEA 20.9 EUR
TCIAB2822-25G TCIAB2822-500G
4-Bromo-2,5-dichlorophenol ≥98.0% (by GC)
4-Bromo-2,5-dichlorophenol
Formule: C₆H₃BrCl₂O
MW: 241.9 g/mol
Kookpunt: 187°C
CAS nummer: 1940-42-7
EINECS: 217-719-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f15e15f69496aef975dbf5164f8fe72b975c2ea4&vid=f15e15f69496aef975dbf5164f8fe72b975c2ea4&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 70.0 to 74.0 deg-C