4-Bromo-3,5-difluoroanisole ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B3392-25G B3392-5G
TCIAB3392-25GEA 256 EUR
TCIAB3392-25G TCIAB3392-5G
4-Bromo-3,5-difluoroanisole ≥98.0% (by GC)
4-Bromo-3,5-difluoroanisole
Formule: C₇H₅BrF₂O
MDL Nummer: MFCD00143268
CAS nummer: 202865-61-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e24b937d3c0de999a5bc375eba1f401d5fc2c6be&vid=e24b937d3c0de999a5bc375eba1f401d5fc2c6be&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR