4-Bromo-3-fluorotoluene ≥94.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
1-Bromo-2-fluoro-4-methylbenzene
B1935-25G
B1935-5G
TCIAB1935-25GEA
94.6
EUR
TCIAB1935-25G
TCIAB1935-5G
4-Bromo-3-fluorotoluene ≥94.0% (by GC)
4-Bromo-3-fluorotoluene
Formule:
C₇H₆BrF MW: 189.03 g/mol Kookpunt: 191°C Vlampunt: 35°C |
MDL Nummer:
MFCD00040940 CAS nummer: 452-74-4 UN: 1993 ADR: 3 III |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 94.0 % |