4-Bromo-2-chlorotoluene ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
1-Bromo-3-chloro-4-methylbenzene
B2068-25G
B2068-5G
TCIAB2068-25GEA
53.9
EUR
TCIAB2068-25G
TCIAB2068-5G
4-Bromo-2-chlorotoluene ≥97.0% (by GC)
4-Bromo-2-chlorotoluene
Formule:
C₇H₆BrCl Kookpunt: 110°C |
MDL Nummer:
MFCD00060651 CAS nummer: 89794-02-5 |
|
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...