2-Bromo-4-chlorotoluene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B3031-25G B3031-5G
TCIAB3031-25GEA 63.8 EUR
TCIAB3031-25G TCIAB3031-5G
2-Bromo-4-chlorotoluene ≥98.0% (by GC)
2-Bromo-4-chlorotoluene
Formule: C₇H₆BrCl
Kookpunt: 114°C
MDL Nummer: MFCD00060650
CAS nummer: 27139-97-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4aecc90dbbd066ce96a962578fc50a5a0d1c51ac&vid=4aecc90dbbd066ce96a962578fc50a5a0d1c51ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR