4-Bromo-2-fluoroanisole ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B1855-25G
B1855-5G
TCIAB1855-25GEA
94.6
EUR
TCIAB1855-25G
TCIAB1855-5G
4-Bromo-2-fluoroanisole ≥98.0% (by GC)
4-Bromo-2-fluoroanisole
Formule:
C₇H₆BrFO Kookpunt: 208°C Smeltpunt: 16 °C Dichtheid: 1,59 g/cm³ (20 °C) Vlampunt: 98°C |
MDL Nummer:
MFCD00011710 CAS nummer: 2357-52-0 EINECS: 219-096-2 |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |