5-Bromo-2-chlorotoluene ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
4-Bromo-1-chloro-2-methylbenzene, 1-Bromo-4-chloro-3-methylbenzene
B3002-5G
B3002-25G
TCIAB3002-5GEA
25.3
EUR
TCIAB3002-5G
TCIAB3002-25G
5-Bromo-2-chlorotoluene ≥98.0% (by GC)
5-Bromo-2-chlorotoluene
Formule:
C₇H₆BrCl MW: 205.48 g/mol Kookpunt: 93°C |
MDL Nummer:
MFCD00018503 CAS nummer: 54932-72-8 |
|
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...