4-Bromo-3-methylanisole ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B1782-25G
B1782-250G
TCIAB1782-25GEA
69.3
EUR
TCIAB1782-25G
TCIAB1782-250G
4-Bromo-3-methylanisole ≥97.0% (by GC)
4-Bromo-3-methylanisole
Formule:
C₈H₉BrO Kookpunt: 135°C |
MDL Nummer:
MFCD00060687 CAS nummer: 27060-75-9 |
|
Specification Test Results
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 97.0 % |
Freezing point | 12.0 to 16.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...