1-Butyl-4-nitrobenzene ≥96.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B3092-25G B3092-5G
TCIAB3092-25GEA 805 EUR
TCIAB3092-25G TCIAB3092-5G
1-Butyl-4-nitrobenzene ≥96.0% (by GC)
1-Butyl-4-nitrobenzene
Formule: C₁₀H₁₃NO₂
Kookpunt: 136°C
CAS nummer: 20651-75-6
EINECS: 243-941-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d22edd2dc62df5ddac93211f042a3c21ff8e42fb&vid=d22edd2dc62df5ddac93211f042a3c21ff8e42fb&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light yellow to Amber to Dark green clear liquid
Purity (GC) min. 96.0 %