2',4',5'-Trifluoroacetophenone ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 1-(2,4,5-Trifluorophenyl)ethan-1-one, 2,4,5-Trifluoroacetophenone

T2758-1G T2758-5G
TCIAT2758-1GEA 17.6 EUR
TCIAT2758-1G TCIAT2758-5G
2',4',5'-Trifluoroacetophenone ≥97.0% (by GC)
2',4',5'-Trifluoroacetophenone
Formule: C₈H₅F₃O
MW: 174.12 g/mol
Kookpunt: 76
Dichtheid: 1.38
Vlampunt: 71°C
MDL Nummer: MFCD00061193
CAS nummer: 129322-83-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83e423a792715128b9e65f3fe5d35ca77a0d45c8&vid=83e423a792715128b9e65f3fe5d35ca77a0d45c8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white clear liquid
Purity (GC) min. 97.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR