3'-Fluoroacetophenone ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

F0262-25G F0262-5G
TCIAF0262-25GEA 112 EUR
TCIAF0262-25G TCIAF0262-5G
3'-Fluoroacetophenone ≥97.0% (by GC)
3'-Fluoroacetophenone
Formule: C₈H₇FO
Kookpunt: 81°C
Smeltpunt: –3 °C
Dichtheid: 1,12 g/cm³ (20 °C)
Vlampunt: 80°C
MDL Nummer: MFCD00000338
CAS nummer: 455-36-7
EINECS: 207-245-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea0dda57f76fc4a9b2a926fc15f035fb70547850&vid=ea0dda57f76fc4a9b2a926fc15f035fb70547850&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR