4'-(Trifluoromethyl)acetophenone ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 1-[4-(Trifluoromethyl)phenyl]ethan-1-one, 4-(Trifluoromethyl)acetophenone, p-(Trifluoromethyl)acetophenone
T1784-1G T1784-25G T1784-5G
TCIAT1784-1GEA 18.7 EUR
TCIAT1784-1G TCIAT1784-25G TCIAT1784-5G
4'-(Trifluoromethyl)acetophenone ≥98.0% (by GC)
4'-(Trifluoromethyl)acetophenone
Formule: C₉H₇F₃O
Kookpunt: 80°C
Smeltpunt: 26…29 °C
Dichtheid: 0,924 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000401
CAS nummer: 709-63-7
EINECS: 211-913-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c41c1802eb7d179d15c6cef2d61ad7f1a0b6c00&vid=5c41c1802eb7d179d15c6cef2d61ad7f1a0b6c00&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %