3-Fluorobenzyl alcohol ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
(3-Fluorophenyl)methanol
F0370-25G
F0370-5G
TCIAF0370-25GEA
123
EUR
TCIAF0370-25G
TCIAF0370-5G
3-Fluorobenzyl alcohol ≥97.0% (by GC)
3-Fluorobenzyl alcohol
Formule:
C₇H₇FO MW: 126.13 g/mol Kookpunt: 201°C Vlampunt: 90°C |
MDL Nummer:
MFCD00004631 CAS nummer: 456-47-3 EINECS: 207-265-3 |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Specific gravity (20/20) | 1.1650 to 1.1750 |
Refractive index n20/D | 1.5080 to 1.5120 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...