5-Fluoro-o-cresol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

F0632-1G F0632-5G
TCIAF0632-1GEA 29.7 EUR
TCIAF0632-1G TCIAF0632-5G
5-Fluoro-o-cresol ≥98.0% (by GC)
5-Fluoro-o-cresol
Formule: C₇H₇FO
Kookpunt: 66°C
MDL Nummer: MFCD00673009
CAS nummer: 452-85-7
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94f6f104ab201a1567e4d2a68dc277e594674ea1&vid=94f6f104ab201a1567e4d2a68dc277e594674ea1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR