3-Fluoro-p-cresol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 3-Fluoro-4-methylphenol, 1-Fluoro-3-hydroxy-6-methylbenzene

F0481-1G F0481-5G
TCIAF0481-1GEA 35.2 EUR
TCIAF0481-1G TCIAF0481-5G
3-Fluoro-p-cresol ≥98.0% (by GC)
3-Fluoro-p-cresol
Formule: C₇H₇FO
Kookpunt: 196°C
MDL Nummer: MFCD00143164
CAS nummer: 452-78-8
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0007bb22068380e93a9775984f9ee59ae2a3117e&vid=0007bb22068380e93a9775984f9ee59ae2a3117e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR