2-Methoxyphenyl isocyanate ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
I0439-25G I0439-5G
TCIAI0439-25GEA 132 EUR
TCIAI0439-25G TCIAI0439-5G
2-Methoxyphenyl isocyanate ≥98.0% (by GC)
2-Methoxyphenyl isocyanate
Formule: C₈H₇NO₂
Kookpunt: 96°C
Dichtheid: 1,122 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00002004
CAS nummer: 700-87-8
EINECS: 211-853-5
UN: 2206
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab3adc86c8c3641dc4b01c60b620d79fa9702441&vid=ab3adc86c8c3641dc4b01c60b620d79fa9702441&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %