1-Propyl isothiocyanate ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
I0388-25ML I0388-5ML
TCIAI0388-25MLEA 99 EUR
TCIAI0388-25ML TCIAI0388-5ML
1-Propyl isothiocyanate ≥98.0% (by GC)
1-Propyl isothiocyanate
Formule: C₄H₇NS
MW: 101.17 g/mol
Kookpunt: 153°C
Dichtheid: 0.9909
Vlampunt: –15°C
MDL Nummer: MFCD00004823
CAS nummer: 628-30-8
EINECS: 211-035-8
UN: 2924
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f086065bb175113421c0f70c30cd818842ccee62&vid=f086065bb175113421c0f70c30cd818842ccee62&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %