4-Methoxythiophenol ≥96.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

M1333-25G M1333-5G
TCIAM1333-25GEA 138 EUR
TCIAM1333-25G TCIAM1333-5G
4-Methoxythiophenol ≥96.0% (by GC)
4-Methoxythiophenol
Formule: C₇H₈OS
Kookpunt: 227°C
Smeltpunt: 42 °C
Dichtheid: 1,1397 g/cm³ (20 °C)
Vlampunt: 96°C
MDL Nummer: MFCD00004849
CAS nummer: 696-63-9
EINECS: 211-799-2
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd835acf17714a69d121bbe4f221c5df4c91fc1d&vid=fd835acf17714a69d121bbe4f221c5df4c91fc1d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR