m-Cymene ≥99.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 3-Methylcumene, 1-Isopropyl-3-methylbenzene, M-cymene, 3-Isopropyltoluene
C0798-0.1ML C0798-1ML
TCIAC0798-0.1MLEA 34.1 EUR
TCIAC0798-0.1ML TCIAC0798-1ML
m-Cymene ≥99.0% (by GC)
m-Cymene
Formule: C₁₀H₁₄
MW: 134.22
Kookpunt: 175
Smeltpunt: -63.8
Dichtheid: 0.865
Vlampunt: 47°C
MDL Nummer: MFCD00008891
CAS nummer: 535-77-3
EINECS: 208-617-9
UN: 2046
ADR: 3 III
Merck Index: 14,02763
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efb33649c957a6634261eafdf1ea02e1cc5c82c7&vid=efb33649c957a6634261eafdf1ea02e1cc5c82c7&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %