p-Cymene ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 1-Isopropyl-4-methylbenzene, 4-Methylcumene, 4-Isopropyltoluene

C0513-25ML C0513-500ML
TCIAC0513-25MLEA 17.6 EUR
TCIAC0513-25ML TCIAC0513-500ML
p-Cymene ≥95.0% (by GC)
p-Cymene
Formule: C₁₀H₁₄
MW: 134.22 g/mol
Kookpunt: 176°C
Smeltpunt: -67.9
Vlampunt: 52°C
MDL Nummer: MFCD00008893
CAS nummer: 99-87-6
EINECS: 202-796-7
UN: 2046
ADR: 3 III
Merck Index: 14,02763
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28424df2791676891ccbec04b9953a1c10f1f481&vid=28424df2791676891ccbec04b9953a1c10f1f481&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR