2,4,5-Trifluorobenzaldehyde ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
T2319-1G T2319-5G
TCIAT2319-1GEA 24.2 EUR
TCIAT2319-1G TCIAT2319-5G
2,4,5-Trifluorobenzaldehyde ≥98.0% (by GC)
2,4,5-Trifluorobenzaldehyde
Formule: C₇H₃F₃O
Kookpunt: 174 °C (1013 hPa)
Dichtheid: 1,408 g/cm³ (20 °C)
Vlampunt: 59°C
MDL Nummer: MFCD00061196
CAS nummer: 165047-24-5
UN: 1989
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc437366df19aae8f54199a250fff23d441ee30c&vid=bc437366df19aae8f54199a250fff23d441ee30c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 98.0 %