2-Bromo-5-chloropyridine ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
B2763-5G B2763-1G
TCIAB2763-5GEA 29.7 EUR
TCIAB2763-5G TCIAB2763-1G
2-Bromo-5-chloropyridine ≥98.0% (by GC)
2-Bromo-5-chloropyridine
Formule: C₅H₃BrClN
Kookpunt: 128°C
MDL Nummer: MFCD00234006
CAS nummer: 40473-01-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=773d83a557320965e6d84e5d1d889fc22a3c8d70&vid=773d83a557320965e6d84e5d1d889fc22a3c8d70&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 65.0 to 69.0 deg-C