2-Bromo-6-fluoropyridine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B3667-1G B3667-5G
TCIAB3667-1GEA 53.9 EUR
TCIAB3667-1G TCIAB3667-5G
2-Bromo-6-fluoropyridine ≥98.0% (by GC)
2-Bromo-6-fluoropyridine
Formule: C₅H₃BrFN
Kookpunt: 164°C
Smeltpunt: 32°C
MDL Nummer: MFCD03095258
CAS nummer: 144100-07-2
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9973262af67b5e56bbd218ac8354b3c54d59f56&vid=e9973262af67b5e56bbd218ac8354b3c54d59f56&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR