6-Methyl-2-pyridone ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
H0942-25G H0942-5G
TCIAH0942-25GEA 101 EUR
TCIAH0942-25G TCIAH0942-5G
6-Methyl-2-pyridone ≥98.0% (by GC, titration analysis)
6-Methyl-2-pyridone
Formule: C₆H₇NO
Kookpunt: 283 - 284 °C (1013 hPa)
Smeltpunt: 159…160 °C
Dichtheid: 1,13 g/cm³ (20 °C)
MDL Nummer: MFCD00006279
CAS nummer: 3279-76-3
EINECS: 221-919-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c37288b09391a2862bbbc0f2bab6b15144f71260&vid=c37288b09391a2862bbbc0f2bab6b15144f71260&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 158.0 to 162.0 deg-C