2-Methyl-4-undecanone ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
I0250-25G I0250-1G
TCIAI0250-25GEA 567 EUR
TCIAI0250-25G TCIAI0250-1G
2-Methyl-4-undecanone ≥97.0% (by GC)
2-Methyl-4-undecanone
Formule: C₁₂H₂₄O
Kookpunt: 124°C
Vlampunt: 91°C
MDL Nummer: MFCD00027319
CAS nummer: 19594-40-2
EINECS: 243-176-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d977fc316ffd3bab4b404231e72c6f0a1027e972&vid=d977fc316ffd3bab4b404231e72c6f0a1027e972&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %