3-Amino-2,6-dichloropyridine ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 2,6-Dichloro-3-pyridinamine, 2,6-Dichloropyridin-3-amine, 2,6-Dichloro-3-pyridylamine
A1580-25G A1580-5G
TCIAA1580-25GEA 224 EUR
TCIAA1580-25G TCIAA1580-5G
3-Amino-2,6-dichloropyridine ≥98.0% (by GC)
3-Amino-2,6-dichloropyridine
Formule: C₅H₄Cl₂N₂
Kookpunt: 305 °C (1013 hPa)
Smeltpunt: 119 °C
Dichtheid: 1,497 g/cm³ (20 °C)
MDL Nummer: MFCD00023417
CAS nummer: 62476-56-6
EINECS: 263-559-1
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7fa2aa706aa620c34b51377c8530b3aa4f176da&vid=c7fa2aa706aa620c34b51377c8530b3aa4f176da&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 119.0 to 123.0 deg-C