3-Bromo-5-fluoropyridine ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
B3244-1G B3244-5G
TCIAB3244-1GEA 25.3 EUR
TCIAB3244-1G TCIAB3244-5G
3-Bromo-5-fluoropyridine ≥98.0% (by GC)
3-Bromo-5-fluoropyridine
Formule: C₅H₃BrFN
MW: 175.99 g/mol
Kookpunt: 78°C
Vlampunt: 64°C
MDL Nummer: MFCD04112555
CAS nummer: 407-20-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=386c6918fa0438e61772a47e71899c1a0b132fab&vid=386c6918fa0438e61772a47e71899c1a0b132fab&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %