3-Bromo-5-methylpyridine ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B3724-1G B3724-5G
TCIAB3724-1GEA 37.4 EUR
TCIAB3724-1G TCIAB3724-5G
3-Bromo-5-methylpyridine ≥98.0% (by GC, titration analysis)
3-Bromo-5-methylpyridine
Formule: C₆H₆BrN
Kookpunt: 110°C
Vlampunt: 91°C
MDL Nummer: MFCD01646141
CAS nummer: 3430-16-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=940478d5b7f425863a7c710cc3983c399b246ae1&vid=940478d5b7f425863a7c710cc3983c399b246ae1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR