5-Bromo-2-methylpyridine ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B2572-1G
B2572-5G
TCIAB2572-1GEA
36.3
EUR
TCIAB2572-1G
TCIAB2572-5G
5-Bromo-2-methylpyridine ≥98.0% (by GC)
5-Bromo-2-methylpyridine
Formule:
C₆H₆BrN Kookpunt: 74°C |
MDL Nummer:
MFCD00661170 CAS nummer: 3430-13-5 |
|
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 34.0 to 38.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...