Propargylalcohol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 2-Propyn-1-ol, Prop-2-yn-1-ol

P0536-25ML P0536-500ML P0536-100ML
TCIAP0536-25MLEA 47.3 EUR
TCIAP0536-25ML TCIAP0536-500ML TCIAP0536-100ML
Propargylalcohol ≥98.0% (by GC)
Propargylalcohol
Formule: C₃H₄O
Kookpunt: 113°C
Smeltpunt: -52°C
Dichtheid: 0,9485 g/cm³ (20 °C)
Vlampunt: 33°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00002912
CAS nummer: 107-19-7
EINECS: 203-471-2
UN: 2929
ADR: 6.1,I
Merck Index: 14,07809
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7a534471bfe39910122a5da60af99c37b9d90f2&vid=d7a534471bfe39910122a5da60af99c37b9d90f2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR