5-Fluoroisatin ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 5-Fluoroindoline-2,3-dione, 5-Fluoro-1H-indole-2,3-dione

F0589-25G F0589-5G
TCIAF0589-25GEA 104 EUR
TCIAF0589-25G TCIAF0589-5G
5-Fluoroisatin ≥98.0% (by GC, titration analysis)
5-Fluoroisatin
Formule: C₈H₄FNO₂
MW: 165.12 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00022795
CAS nummer: 443-69-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fa8b357345e76ecbe85eb6436f4bd73a713952d&vid=5fa8b357345e76ecbe85eb6436f4bd73a713952d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 224.0 to 230.0 deg-C

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR