4-Benzoylbenzoic acid ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B1164-25G B1164-5G
TCIAB1164-25GEA 213 EUR
TCIAB1164-25G TCIAB1164-5G
4-Benzoylbenzoic acid ≥98.0% (by titrimetric analysis)
4-Benzoylbenzoic acid
Formule: C₁₄H₁₀O₃
Kookpunt: 416 °C (1013 hPa)
Smeltpunt: 69 °C
MDL Nummer: MFCD00002560
CAS nummer: 611-95-0
EINECS: 210-286-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3187189e8093aff72fe8cacfb7cca39d6e3e3b15&vid=3187189e8093aff72fe8cacfb7cca39d6e3e3b15&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 196.0 to 202.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR