4-Chlorobenzamide ≥98.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

C0563-25G C0563-250G
TCIAC0563-25GEA 89.1 EUR
TCIAC0563-25G TCIAC0563-250G
4-Chlorobenzamide ≥98.0% (by HPLC, total nitrogen)
4-Chlorobenzamide
Formule: C₇H₆ClNO
Kookpunt: 314 °C (1013 hPa)
Smeltpunt: 179 °C
MDL Nummer: MFCD00007993
CAS nummer: 619-56-7
EINECS: 210-601-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=847b52a36a23e39b2233efd3be213750731e4efb&vid=847b52a36a23e39b2233efd3be213750731e4efb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 179.0 to 182.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR