4-Acetylbenzoic acid ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

A1024-1G A1024-5G
TCIAA1024-1GEA 22 EUR
TCIAA1024-1G TCIAA1024-5G
4-Acetylbenzoic acid ≥98.0% (by titrimetric analysis)
4-Acetylbenzoic acid
Formule: C₉H₈O₃
Kookpunt: 340 °C (1013 hPa)
MDL Nummer: MFCD00002561
CAS nummer: 586-89-0
EINECS: 209-588-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff8855a2a96113fd2c4802b022cde862601694ff&vid=ff8855a2a96113fd2c4802b022cde862601694ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 208.0 to 212.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR