4-Heptylbenzoyl chloride ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
H0792-25G
H0792-5G
TCIAH0792-25GEA
720
EUR
TCIAH0792-25G
TCIAH0792-5G
4-Heptylbenzoyl chloride ≥98.0% (by titrimetric analysis)
4-Heptylbenzoyl chloride
Formule:
C₁₄H₁₉ClO Kookpunt: 196°C Dichtheid: 1,002 g/cm³ (20 °C) |
MDL Nummer:
MFCD00000702 CAS nummer: 50606-96-7 EINECS: 256-648-1 UN: 3265 ADR: 8,III |
|
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...