4-Iodobenzoyl chloride ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: p-Iodobenzoyl chloride
I0508-25G I0508-5G
TCIAI0508-25GEA 169 EUR
TCIAI0508-25G TCIAI0508-5G
4-Iodobenzoyl chloride ≥98.0% (by GC, titration analysis)
4-Iodobenzoyl chloride
Formule: C₇H₄ClIO
MW: 266.47 g/mol
Kookpunt: 100°C
MDL Nummer: MFCD00001058
CAS nummer: 1711-02-0
EINECS: 216-974-7
UN: 3261
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7618a56308f9bedde138b687d1c55e5d49867192&vid=7618a56308f9bedde138b687d1c55e5d49867192&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 63.0 to 67.0 deg-C