5-Bromoindole ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 5-Bromo-1H-indole

B1738-25G B1738-5G
TCIAB1738-25GEA 71.5 EUR
TCIAB1738-25G TCIAB1738-5G
5-Bromoindole ≥99.0% (by GC)
5-Bromoindole
Formule: C₈H₆BrN
MW: 196.05 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005670
CAS nummer: 10075-50-0
EINECS: 233-208-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33e24d29fe7a7a517493e21ae2da4845393cf6a2&vid=33e24d29fe7a7a517493e21ae2da4845393cf6a2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 99.0 %
Melting point 88.0 to 92.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR